Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences | Avantor
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Certificates
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All of our Dullbecco’s phosphate buffer saline solutions contain: potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose.
  • Some formulations may contain additional components
  • Store at room temperature
Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.
Product Family Options
Informations sur le produit
  • Désignation
    Concentration
    Taille
    Type de conditionnement
    Availability
    Price
  • D-PBS, Formula I Phosphate Buffer Saline
    0.15 M
    500 ml
    Bottle
    Discontinued
    Supplier Number: 11772-05
  • D-PBS, Formula I Phosphate Buffer Saline
    0.15 M
    1 l
    Bottle
    Discontinued
    Supplier Number: 11772-10
  • D-PBS, Formula II Phosphate Buffer Saline
    0.19 M
    500 ml
    Bottle
    Discontinued
    Supplier Number: 11773-05
  • D-PBS, Formula II Phosphate Buffer Saline
    0.19 M
    1 l
    Bottle
    Discontinued
    Supplier Number: 11773-10
  • D-PBS, Formula III with phenol red, Phosphate Buffer Saline
    0.15 M
    500 ml
    Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500ml)
    Each (500ml) - $90.02
    $90.02
    Each (500ml)
       $90.02
    Supplier Number: 11774-05
  • D-PBS, Formula III with phenol red, Phosphate Buffer Saline
    0.15 M
    1 l
    Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.60
    $103.60
    Each
       $103.60
    Supplier Number: 11774-10