Millonig's Phosphate Buffers, Electron Microscopy Sciences
Millonig's Phosphate Buffers, Electron Microscopy Sciences
Millonig's Phosphate Buffers, Electron Microscopy Sciences
Millonig's Phosphate Buffers, Electron Microscopy Sciences
Millonig's Phosphate Buffers, Electron Microscopy Sciences
Millonig recommended to use this buffers for the fixation of very hydrated tissues with osmium tetroxide. For marine organisms, a higher concentration of sodium chloride (3%) is recommended.
  • Ingredients: Sodium phosphate, monobasic (NaH2PO4.xH2O), sodium phosphate, dibasic (Na2HPO4.xH2O), 0.5% sodium chloride (NaCl)
The pH of this buffers are 7.4 (0.1 M) and the osmolarity is 440 mosmols, so this buffers are hypertonic to most body fluids.
Product Family Options
  • Désignation
    Type de conditionnement
    Taille
    pH
    Température de stockage
  • Millonig's Phosphate Buffer, 0.2 M
    Bottle
    500 ml
    7.4 (0.1 M)
    0...5 °C
    Each (500ml)
    Each (500ml) - $69.70
    $69.70
    Each (500ml)
       $69.70
     : 11582-05
  • Millonig's Phosphate Buffer, 0.2 M
    Bottle
    1 l
    7.4 (0.1 M)
    0...5 °C
    Each
    Each - $98.41
    $98.41
    Each
       $98.41
     : 11582-10