Hollande's Fixative, Electron Microscopy Science
À propos de cet article
Hollande's fixative is a modification of Bouin's solution. Used as a fixative for biopsy specimens of the gastrointestinal tract.
- Stable and will decalcify small bone specimens
- Prefilled ready-to-use
A modification of Bouin's solution. It is stable and will decalcify small bone specimens. Tissue that is fixed with Hollande's can be stained successfully with most stains, and the cupric acetate in the solution stabilizes red blood cell membranes and cosinophil and endocrine cell granules so that less lysis occurs than with Bouin's solution. Hollande's is widely used as a fixative for biopsy specimens of the gastrointestinal tract.
Thorough washing of the fixative prior to placing the specimen in a phosphate buffered formalin solution is necessary because the salts present in the solution will form an insoluble phosphate precipitate. Packaged in leak-proof histo-containers; prefilled ready-to-use.
: Packaged in leak-proof histo-containers.
1 Options Available Below
Product Details & Documents
Hollande's fixative is a modification of Bouin's solution. Used as a fixative for biopsy specimens of the gastrointestinal tract.
- Stable and will decalcify small bone specimens
- Prefilled ready-to-use
A modification of Bouin's solution. It is stable and will decalcify small bone specimens. Tissue that is fixed with Hollande's can be stained successfully with most stains, and the cupric acetate in the solution stabilizes red blood cell membranes and cosinophil and endocrine cell granules so that less lysis occurs than with Bouin's solution. Hollande's is widely used as a fixative for biopsy specimens of the gastrointestinal tract.
Thorough washing of the fixative prior to placing the specimen in a phosphate buffered formalin solution is necessary because the salts present in the solution will form an insoluble phosphate precipitate. Packaged in leak-proof histo-containers; prefilled ready-to-use.
: Packaged in leak-proof histo-containers.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 100502-582
- 100500-432
- 100498-614
- 100498-616
- Boiling point:
- -21 °C (-6 °F)
- UN No.:
- 1760
- Storage temperature:
- RT
Spécifications
Product Family Options
Informations sur le produit
- DésignationTailleAvailabilityPrice
- Hollande's Fixative20 mlCase of 50$280.62Each$280.62Supplier Number: 64120-20Spécifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100502-582Boiling pointUN No.Storage temperatureMinority or Woman-Owned Business Enterprise - Hollande's Fixative30 mlDiscontinued-Supplier Number: 64120-30Spécifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100500-432Boiling pointUN No.Storage temperatureMinority or Woman-Owned Business Enterprise - Hollande's Fixative1 LDiscontinued-Supplier Number: 64120-01Spécifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100498-614Boiling point-21 °C (-6 °F)UN No.1760Storage temperatureRTMinority or Woman-Owned Business Enterprise - Hollande's Fixative4 LDiscontinued-Supplier Number: 64120-04Spécifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100498-616Boiling pointUN No.Storage temperatureMinority or Woman-Owned Business Enterprise