HANKS's Balanced Salts, Electron Microscopy Sciences | Avantor
HANKS's Balanced Salts, Electron Microscopy Sciences
HANKS's Balanced Salts, Electron Microscopy Sciences
Certificates
HANKS's Balanced Salts, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All of our Hanks’ Balanced Salt Solutions contain: Potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application.
All solutions are prepared in 1X strength.
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • 100502-390
  • 100502-388
  • 100496-428
  • 100502-386
  • Size:
  • 500 ml
  • 500 ml
  • 1 L
  • 500 ml
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Formula III
    Discontinued
    Supplier Number: 11770-05
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100502-390
    Size
    500 ml
    Minority or Woman-Owned Business Enterprise
  • Formula II
    Discontinued
    Supplier Number: 11769-05
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100502-388
    Size
    500 ml
    Minority or Woman-Owned Business Enterprise
  • Formula III
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1l)
    Each (1l) - $87.11
    $87.11
    Each (1l)
       $87.11
    Supplier Number: 11770-10
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100496-428
    Size
    1 L
    Minority or Woman-Owned Business Enterprise
  • 500 ml
    Discontinued
    Supplier Number: 11768-05
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100502-386
    Size
    500 ml
    Minority or Woman-Owned Business Enterprise