eShield™ Sterile Tablet Cover, Whitney Medical Solutions
10798-988
- Description:Sterile tablet cover
- Material:Ultra-clear polyethylene
- Cat. no.:10798-988
- Supplier No.:EC2300
eShield™ sterile tablet covers are designed to protect a sterile environment from contaminants on the device.
- Covers are touchscreen compatible, even with double gloves
- Tablet covers are designed to accommodate aseptic technique
- Constructed of an ultra-clear polyethylene for clear photos
The sterile barrier provided by eShield™ enables the medical professional to access tablets to utilize medical apps, access studies, record data, conduct consultations, and capture photos conveniently and safely without contaminating the sterile environment.
They can be used within the sterile field in any healthcare environment, including surgical suites and clinical and research labs.
: FDA Approved. Meets ASTM F1671 Viral penetration guidelines.
: Contact your Avantor sales representative for more information.