Cryogenic Printable Labels, Brady
À propos de cet article
Cryogenic labels for tubes, vials and canes.
Chemical and stain resistant labels for slides and cassettes:
Temperatures from –80 to 121 °C (–112 to 250 °F).
B-449 Slide label with removability and write on capabilities.
B-481 Stainerbondz™ extreme chemical resistant slide label.
B-482 Tissue cassette label applied pre-process with the BSP®31 for extreme solvent/stain resistance.
B-488 Slide label with moderate chemical.
B-498 Tissue cassette label for re-identification of processed paraffin blocks.
General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more:
B-424 Economical paper label for general purpose.
B-427 Straw label for wrapping; working temperatures from –196 to 80 °C (–321 to 176 °F).
B-430 Petri dish and bag label; working temperatures from –80 to 121 °C (–112 to 250 °F).
B-499 For cryo boxes, freezer racks, petri dishes, bottles, flasks, microplates, and bags; working temperatures from –196 to 121 °C (–321 to 250 °F).
41 Options Available Below
Product Details & Documents
Cryogenic labels for tubes, vials and canes.
Chemical and stain resistant labels for slides and cassettes:
Temperatures from –80 to 121 °C (–112 to 250 °F).
B-449 Slide label with removability and write on capabilities.
B-481 Stainerbondz™ extreme chemical resistant slide label.
B-482 Tissue cassette label applied pre-process with the BSP®31 for extreme solvent/stain resistance.
B-488 Slide label with moderate chemical.
B-498 Tissue cassette label for re-identification of processed paraffin blocks.
General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more:
B-424 Economical paper label for general purpose.
B-427 Straw label for wrapping; working temperatures from –196 to 80 °C (–321 to 176 °F).
B-430 Petri dish and bag label; working temperatures from –80 to 121 °C (–112 to 250 °F).
B-499 For cryo boxes, freezer racks, petri dishes, bottles, flasks, microplates, and bags; working temperatures from –196 to 121 °C (–321 to 250 °F).
Product Family Options
B-427 Straw Label for Wrapping
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Thermal transfer labelsVinylWhite/translucentPR300+, PR600+, IP300®, IP600®, thermal transfer printers3.75×1" (1×1" printable area)Log in pour la disponibilitéEach$853.61Supplier Number: THT-10-427-3
- 1 - 2 ml vial labels for thermal printerVinylWhite/translucentPR300+, PR600+, IP300®, IP600®, thermal transfer printers1×2.625"Log in pour la disponibilitéEachEach - $1,142.34$1,142.34Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $1,142.34$1,142.34Prix promotionnel pour les nouveaux clientsEach$1,142.34Roll of 3,000$1,142.34Supplier Number: THT-131-427-3
B-499 For Cryo Boxes, Freezer Racks, Petri Dishes, Bottles, Flasks, Microplates, and Bags
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Thermal transer labelsNylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers1.44×0.8"Log in pour la disponibilitéEach$400.37Supplier Number: THT-69-499-3
- Thermal transfer labelsNylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers0.375×1"Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $427.02$427.02Prix promotionnel pour les nouveaux clientsBox of 3,000$427.02Supplier Number: THT-152-499-3
- Nylon cloth label b499Nylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers0.375"Log in pour la disponibilitéRoll of 3,000Roll of 3,000 - $447.82$447.82Prix promotionnel pour les nouveaux clientsEach$447.82Roll of 3,000$447.82Supplier Number: THT-163-499-3
- Thermal transfer labels for 0.6 ml vials and tubesNylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers0.5×0.9"Discontinued-Supplier Number: -
- Thermal transfer label for 1.5 to 2 ml tubesNylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers0.5"Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $692.47$692.47Prix promotionnel pour les nouveaux clientsBox of 3,000$692.47Supplier Number: THT-126-499-3
- Thermal transfer labels for 50 ml conical tubesNylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers1×2"Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $1,424.13$1,424.13Prix promotionnel pour les nouveaux clientsBox of 3,000$1,424.13Supplier Number: THT-137-499-3
- Thermal transfer labels, nylon cloth, 1.5"Nylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers1.5×0.75"Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $809.09$809.09Prix promotionnel pour les nouveaux clientsBox of 3,000$809.09Supplier Number: THT-136-499-3
- Thermal transfer printer labelsNylon ClothSilverBBP®81, IP® series, PR Plus series1.75×1.875"Log in pour la disponibilitéEachEach - $1,796.39$1,796.39Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéRoll of 1Roll of 1 - $1,796.39$1,796.39Prix promotionnel pour les nouveaux clientsEach$1,796.39Roll of 1$1,796.39Supplier Number: 104790
- Microplates application labels, B499Nylon ClothWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers2×0.25"Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $580.48$580.48Prix promotionnel pour les nouveaux clientsBox of 3,000$580.48Supplier Number: THT-151-499-3
B-351 Removes in Fragments, Good Resistance to Solvents and Humidity, UL and RoHS
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Tht printer labels B-351 MAT´LVinylWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers0.5×1"Log in pour la disponibilitéEach$917.52Supplier Number: THT-5-351-10
- Tht printer labels B-351 MAT´LVinylWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers1×2"Log in pour la disponibilitéEach$575.36Supplier Number: THT-17-351-3
B-461 Ultra-Thin Label with Clear Tail for Tubes
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Self laminating labelsPolyesterWhite/translucentIP300®, IP600®, BBP® 81, thermal transfer printers41.3×15.2 mm (1.63×0.6")Discontinued-Supplier Number: THT-133-461-3
- Self laminating labelsPolyesterWhite/translucentIP300®, IP600®, BBP® 81, thermal transfer printers45.7×12.7 mm (1.8×0.5")Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $478.96$478.96Prix promotionnel pour les nouveaux clientsBox of 3,000$478.96Supplier Number: THT-122-461-3
- Thermal transfer printable labelsPolyesterWhite/translucentIP300®, IP600®, BBP® 81, thermal transfer printers45.7×9.7 mm (1.8×0.38")Log in pour la disponibilitéBox of 3,000Box of 3,000 - $314.36$314.36Prix promotionnel pour les nouveaux clientsBox of 3,000$314.36Each$314.36Supplier Number: THT-123-461-3
- Self laminating labelsPolyesterWhite/translucentIP300®, IP600®, BBP® 81, thermal transfer printers55.9×12.7 mm (2.2×0.5")Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $577.08$577.08Prix promotionnel pour les nouveaux clientsBox of 3,000$577.08Supplier Number: THT-127-461-3
- Thermal transfer printable labelsPolyesterWhite/translucentIP300®, IP600®, BBP® 81, thermal transfer printers133.4×25.4 mm (5.25×1")Log in pour la disponibilitéEachEach - $1,048.94$1,048.94Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéRoll of 2,000Roll of 2,000 - $1,048.94$1,048.94Prix promotionnel pour les nouveaux clientsEach$1,048.94Roll of 2,000$1,048.94Supplier Number: THT-139-461-2
B-423 Product and Component Identification Labels, PCB and Rating Plates
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Discontinued-Supplier Number: -
- Cryogenic printable labelsPolyesterWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers0.25×2"2500 labels/rollLog in pour la disponibilitéRoll of 2,500Roll of 2,500 - $134.56$134.56Prix promotionnel pour les nouveaux clientsEach$134.56Roll of 2,500$134.56Supplier Number: THT-15-423-2.5
- Cryogenic printable labelsPolyesterWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers0.25×2"10000 labels/boxLog in pour la disponibilitéBox of 10,000Box of 10,000 - $596.94$596.94Prix promotionnel pour les nouveaux clientsBox of 10,000$596.94Each$596.94Supplier Number: 30513
B-490 Freezerbondz® Label for Frozen Tubes
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Cryogenic printable labelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers15.2×41.4 mm (0.6×1.63")Discontinued-Supplier Number: THT-155-490-3
- Cryogenic printable labelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers25.4×48.3 mm (1×1.9")Log in pour la disponibilitéEachEach - $1,013.25$1,013.25Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $1,013.25$1,013.25Prix promotionnel pour les nouveaux clientsEach$1,013.25Roll of 3,000$1,013.25Supplier Number: THT-156-490-3
- Cryogenic printable labelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers38.1×55.9 mm (1.5×2.2")Log in pour la disponibilitéEachEach - $1,469.54$1,469.54Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $1,469.54$1,469.54Prix promotionnel pour les nouveaux clientsEach$1,469.54Roll of 3,000$1,469.54Supplier Number: THT-157-490-3
Ribbon
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Discontinued-Supplier Number: -
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers1"984 ft.Log in pour la disponibilitéEach$183.21Supplier Number: 10615
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers1.57"984 ft.Log in pour la disponibilitéEach$319.18Supplier Number: 66261
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers1.57"984 ft.Log in pour la disponibilitéEach$324.87Supplier Number: IP-R6406
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers2.36"984 ft.Log in pour la disponibilitéEach$298.15Supplier Number: 54802
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers2.36"984 ft.Log in pour la disponibilitéEach$294.52Supplier Number: 35269
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers2.36"984 ft.Log in pour la disponibilitéEach$461.20Supplier Number: 66263
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers3.27"984 ft.Log in pour la disponibilitéEach$657.30Supplier Number: 110380
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers4"360 ft.Log in pour la disponibilitéEach$275.84Supplier Number: 11510
- Cryogenic printable labelsBlackPR300+, PR600+, IP300®, IP600®, thermal transfer printers4.33"984 ft.Log in pour la disponibilitéEach$896.69Supplier Number: 110381
B-492 Freezerbondz® Ultra-Thin Label for Frosted/Frozen Tubes and Canes
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Freezerbondz II, B492 labelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.375×1"Discontinued-Supplier Number: THT-152-492-3
- Freezerbondz II, B492PolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.5×1"Discontinued-Supplier Number: -
- LabelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.5"Log in pour la disponibilitéLog in pour la disponibilitéBox of 3,000Box of 3,000 - $515.25$515.25Prix promotionnel pour les nouveaux clientsBox of 3,000$515.25Supplier Number: THT-144-492-3
- Freezerbondz II, B492 labelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers1×1"Log in pour la disponibilitéRoll of 3,000Roll of 3,000 - $877.25$877.25Prix promotionnel pour les nouveaux clientsEach$877.25Roll of 3,000$877.25Supplier Number: THT-179-492-3
- Freezerbondz™ thermal transfer printer labelsPolyesterWhitePR300+, PR600+, IP300®, IP600®, thermal transfer printers1×1.9"Discontinued-Supplier Number: -
- Freezerbondz II, B492 labelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.375"Discontinued-Supplier Number: -
- Freezerbondz II, B492 labelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.375"Discontinued-Supplier Number: -
- Freezerbondz thermal transfer printer labels, 3000/rollPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.375"Discontinued-Supplier Number: -
B-449 Slide Label with Removability and Write-On Capabilities
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- PP labelsPPWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.75×1.5"Log in pour la disponibilitéLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $289.73$289.73Prix promotionnel pour les nouveaux clientsRoll of 3,000$289.73Supplier Number: 116392
B-481 Stainerbondz™ Extreme Chemical Resistant Slide Label
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Stainerbondz, b481, slides applicationPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.75×0.9"Log in pour la disponibilitéRoll of 3,000Roll of 3,000 - $763.85$763.85Prix promotionnel pour les nouveaux clientsEach$763.85Roll of 3,000$763.85Supplier Number: THT-194-481-3
- Slide labels, chemical resis., 3000/rollPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.9×0.9"Discontinued-Supplier Number: -
- LabelsPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers2.5×2.5"Log in pour la disponibilitéLog in pour la disponibilitéRoll of 2,500Roll of 2,500 - $1,163.73$1,163.73Prix promotionnel pour les nouveaux clientsRoll of 2,500$1,163.73Supplier Number: 116409
B-488 Slide Label with Moderate Chemical Resistance
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- High performance, B488, slides applicationPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.9×0.75"Discontinued-Supplier Number: -
- B488, slides applicationPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.9×0.9"Log in pour la disponibilitéRoll of 3,000Roll of 3,000 - $314.58$314.58Prix promotionnel pour les nouveaux clientsEach$314.58Roll of 3,000$314.58Supplier Number: THT-141-488-3
- B488 matte slide labelPolyesterWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.9×0.9"Log in pour la disponibilitéRoll of 1,000Roll of 1,000 - $131.48$131.48Prix promotionnel pour les nouveaux clientsEach$131.48Roll of 1,000$131.48Supplier Number: THT-164-488-1
B-498 Tissue Cassette Label for Re-identification of Processed Paraffin Blocks
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDiameterWidthDimensionsLongueurRoll LengthPackagingAvailabilityPrice
- Vinyl labelsVinyl ClothWhiteIP300®, IP600®, BBP®81, thermal transfer printers0.25×15"Log in pour la disponibilitéLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $592.92$592.92Prix promotionnel pour les nouveaux clientsRoll of 3,000$592.92Supplier Number: 116407
- Vinyl labelsVinyl ClothWhiteBBP®81, IP300®, IP600®, PR300+, PR600+1"0.250.75"Log in pour la disponibilitéLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $719.90$719.90Prix promotionnel pour les nouveaux clientsRoll of 3,000$719.90Supplier Number: 142210
- Vinyl labelsVinyl ClothWhiteBBP®81, IP300®, IP600®, PR300+, PR600+1"0.75"0.75"Log in pour la disponibilitéLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $715.62$715.62Prix promotionnel pour les nouveaux clientsRoll of 3,000$715.62Supplier Number: 142212
- Vinyl cloth labelsVinyl ClothWhiteIP300®, IP600®, BBP®81, thermal transfer printers2×1"Log in pour la disponibilitéLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $2,593.58$2,593.58Prix promotionnel pour les nouveaux clientsRoll of 3,000$2,593.58Supplier Number: 116413
- Vinyl labelsVinyl ClothWhiteBBP®81, IP300®, IP600®, PR300+, PR600+Log in pour la disponibilitéLog in pour la disponibilitéRoll of 3,000Roll of 3,000 - $721.09$721.09Prix promotionnel pour les nouveaux clientsRoll of 3,000$721.09Supplier Number: 142211