To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Length:241 mm (9½")
- Material:Stainless Steel with Reduced Reflectivity Polymer Coating
- Tip Configuration:1 x 2 Nontraumatic Fine Teeth
- Cat. no.:10199-116
- Tip Style:Straight, Blunt
- Supplier No.:90-2312
Sklar Black™ DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps, used for grasping fine tissue during laser surgeries.
It is available in multiple lengths, straight or angled, and with a variety of tip dimensions. They are commonly used in vascular and general surgery.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.