ContactUsLinkComponent
Sklar Black™ DeBakey Forceps, OR Grade, Sklar
  10199-116
 :  Sklar
undefined
Sklar Black™ DeBakey Forceps, OR Grade, Sklar
  10199-116
 :  Sklar
 :  90-2312
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Length:
    241 mm (9½")
  • Material:
    Stainless Steel with Reduced Reflectivity Polymer Coating
  • Tip Configuration:
    1 x 2 Nontraumatic Fine Teeth
  • Cat. no.:
    10199-116
  • Tip Style:
    Straight, Blunt
  • Supplier No.:
    90-2312
Sklar Black™ DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps, used for grasping fine tissue during laser surgeries.
It is available in multiple lengths, straight or angled, and with a variety of tip dimensions. They are commonly used in vascular and general surgery.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
 : All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.