Micro Scissors, Sklar
Micro Scissors, Sklar
Supplier:   Sklar
Micro Scissors, Sklar
Supplier:  Sklar
Certificates
Micro Scissors, Sklar
Micro Scissors, Sklar
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sklar's® micro scissors are used for the dissection of tissues, blood vessels and nerves.
  • Reusable
  • Nonsterile
Available with a flat or round handle. The straight scissors may be used to cut sutures and trim nerve endings. Curved will dissect vessels and nerves. In addition to the tips, the sides may be used for dissection with the tips closed, similar to using forceps.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Type
    Curvature
    Availability
    Price
  • Micro Scissors, Round Handle
    152 mm (6")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,454.73
    $1,454.73
    Prix promotionnel pour les nouveaux clients
    Each
       $1,454.73
    Supplier Number: 98-7814
  • Micro Scissors, Flat Handle
    178 mm (7")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,546.26
    $1,546.26
    Prix promotionnel pour les nouveaux clients
    Each
       $1,546.26
    Supplier Number: 98-7813
  • Micro Scissors, Round Handle
    178 mm (7")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,964.15
    $2,964.15
    Prix promotionnel pour les nouveaux clients
    Each
       $2,964.15
    Supplier Number: 98-7837
  • Micro Scissors, Flat Handle
    203 mm (8")
    Stainless steel
    Sharp/Sharp, Serrated
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,007.58
    $3,007.58
    Prix promotionnel pour les nouveaux clients
    Each
       $3,007.58
    Supplier Number: 98-7892