Hartmann Alligator Ear Forceps, OR Grade, Sklar
Supplier: Sklar
Certificates
About this item
Hartmann Alligator Forceps are an ENT instrument used in myringotomy procedures to insert or remove tubes or dressings and packing.
- This product has 3.5" shanks that angle at a screw lock to a 2.5" shaft and 8mm long triangular tips. The tips are horizontally serrated and each have a central groove to securely grip surgical materials, as well as granulation tissue and cerumen.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together