Barrett Tenaculum Forceps, OR Grade, Sklar
Supplier:   Sklar
Barrett Tenaculum Forceps, OR Grade, Sklar
Supplier:  Sklar
Certificates
Barrett Tenaculum Forceps, OR Grade, Sklar
Barrett Tenaculum Forceps, OR Grade, Sklar
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Barrett Tenaculum Forceps are ratcheted, finger ring forceps used for grasping cervical tissue.
With straight shanks and sharp tips that curve inward, Barrett tenaculum forceps resemble Braun tenaculum forceps; however, Barrett forceps are shorter
Censitrac Ready products are industry standard and constructed of stainless steel.
The sharp, pointed tips ensure a firm grip on the targeted tissue. These forceps may be used for foreign body removal.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Product Family Options
Informations sur le produit
  • Désignation
    Grade
    Matière
    Longueur
    Tip Style
    Handle
    Tip Configuration
    Curvature
    Sterile
    Availability
    Price
  • Barrett Tenaculum Forceps, Delicate
    Premium OR-Grade
    Stainless steel
    16.5 cm (6.5")
    Pointed
    Finger Rings
    1 x 1 Prongs Sharp
    Straight
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 590,48 $
    590,48 $
    Prix promotionnel pour les nouveaux clients
    Each
       590,48 $
    Supplier Number: 18-2257
  • Barrett Tenaculum Forceps, Delicate
    Premium OR
    Stainless steel
    17.8 cm (7")
    Pointed
    Finger Rings
    1 x 1 Prongs
    Straight
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 397,43 $
    397,43 $
    Prix promotionnel pour les nouveaux clients
    Each
       397,43 $
    Supplier Number: 91-1070