PC Link Labels, Brady
B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
B-499: Nylon cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
B-423: Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-351: Tamper-resistant vinyl is designed to fracture easily to show signs of product tampering and to prevent on-piece label removal.
B-435: Glossy metallized permanent polyester is designed for high adhesion to textured metals, powder coated surfaces and low surface energy plastics.
B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.
B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
B-499: Nylon cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
B-423: Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-351: Tamper-resistant vinyl is designed to fracture easily to show signs of product tampering and to prevent on-piece label removal.
B-435: Glossy metallized permanent polyester is designed for high adhesion to textured metals, powder coated surfaces and low surface energy plastics.
B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.
Product Family Options
B-461 Ultra-Thin Label with Clear Tail for Tubes
- DésignationMatièreColorPour l'utilisation avecWidthDiameterDimensionsRoll Length
- Tls printer labels B-461 MAT´LPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®610.75×3.75"Each$61.14
Specifications
Cat. No.CA80090-148 - PCR tube labelsPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®610.19×1.5"-
Specifications
Cat. No.490006-206 - Polyester labelsPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®610.375×1.8"-
Specifications
Cat. No.490006-226 - Self laminating labelsPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®610.5×1.8"-
Specifications
Cat. No.490006-224 - Labels for portable thermal printingPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®610.5×2.2"-
Specifications
Cat. No.490006-228 - Polyester labels for 1-2 ml tubePolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®610.6×1.625"-
Specifications
Cat. No.490006-234 - Labels for portable thermal printingPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®610.75×3.75"-
Specifications
Cat. No.490006-246 - Polyester labels for 1-8 ml vialPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®611×2.625"-
Specifications
Cat. No.490006-232 - Polyester labels for 15 ml tubePolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®611.5×3.5"-
Specifications
Cat. No.490006-238 - Polyester labels for vialsPolyesterWhite/translucentTLS 2200®, TLS PC Link®, BMP®71, BMP®612.625×0.6"-
Specifications
Cat. No.490006-262
Other TLS 2200®/TLS PC Link® Ribbons
- DésignationMatièreColorPour l'utilisation avecWidthDiameterDimensionsRoll Length
- : -
Specifications
Cat. No.CA80079-942 - : R4410-WT
Specifications
Cat. No.CA80090-200 - Tls print ribbon redRedTLS 2200®, TLS PC Link®2"75 ft.-: -
Specifications
Cat. No.CA80090-202 - RibbonsBlackTLS 2200®, TLS PC Link®2"75 ft.-: -
Specifications
Cat. No.490006-286 - RibbonsBlackTLS 2200®, TLS PC Link®2"75 ft.-: -
Specifications
Cat. No.490006-280 - RibbonsBlackTLS 2200®, TLS PC Link®2"75 ft.-: -
Specifications
Cat. No.490006-272