Angular Scissors, OR Grade, Sklar
Product Details & Documents
Angular Scissors are commonly used in dental and ophthalmologic procedures.
The angled blades are smooth and have sharp/sharp tips.
Angular Scissors are finger ring scissors comparable to iris scissors.
Spécifications
- Longueur:121 mm (4¾")
- Matière:Stainless steel
- Blade Type:Angled, Smooth, Sharp/Sharp