pH Buffer Solutions, Hach
pH Buffer Solutions, Hach
 :   Hach
pH Buffer Solutions, Hach
 :  Hach
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach
For pH meter standardization
Available colorless, or color-coded for easy identification.
Documents
  • Catalog No:
  • 10199-502
  • CA10199-502
  • 10199-498
  • 10199-500
  • CA10199-500
  • CA97021-520
  • CA22834-56
  • CA97021-444
  • 10199-506
  • CA10199-506
  • CA11017-014
  • 10199-504
  • CA10199-504
  • CA97021-458
  • CA97021-426
  • CA97021-440
  • CA97021-432
  • 10199-510
  • CA10199-510
  • 10199-508
  • CA10199-508
  • CA97021-552
  • CA97021-408
  • CA22836-56
  • CA10199-498
  • Concentration:
  • 10.01 pH ± 0.02 pH at 25 °C
  • 10.01 pH
  • 10.01 ± 0.02 pH
Product Family Options
  • pH Accuracy
    Reference Temperature
    Taille
    pH
    Color
  • ±0.02
    25 °C
    15 mL
    4.01
    Red
     : -
    Specifications
    Cat. No.
    10199-502
    Concentration
  • ±0.02
    25 °C
    15 mL
    4.01
    Red
    Each
    Each - $23.65
    $23.65
    Each
       $23.65
     : 2283436
    Specifications
    Cat. No.
    CA10199-502
    Concentration
  • ±0.02
    25 °C
    15 mL
    4.01
    Colorless
     : -
    Specifications
    Cat. No.
    10199-498
    Concentration
  • ±0.02
    25 °C
    50 mL
    4.01
    Red
     : -
    Specifications
    Cat. No.
    10199-500
    Concentration
  • ±0.02
    25 °C
    50 mL
    4.01
    Red
    Each
    Each - $34.77
    $34.77
    Each
       $34.77
     : 2283426
    Specifications
    Cat. No.
    CA10199-500
    Concentration
  • ±0.02
    25 °C
    500 mL
    4.01
    Red
    Each (500ml)
    Each (500ml) - $57.40
    $57.40
    Each (500ml)
       $57.40
     : 2283449
    Specifications
    Cat. No.
    CA97021-520
    Concentration
  • ±0.02
    25 °C
    4 L
    4.01
    Red
    Each (4l)
    Each (4l) - $174.03
    $174.03
    Each (4l)
       $174.03
     : 2283456
    Specifications
    Cat. No.
    CA22834-56
    Concentration
  • ±0.02
    25 °C
    4 L
    4.01
    Colorless
    Each (4l)
    Each (4l) - $208.70
    $208.70
    Each (4l)
       $208.70
     : 1222356
    Specifications
    Cat. No.
    CA97021-444
    Concentration
  • ±0.02
    25 °C
    15 mL
    7.00
    Yellow
     : 2283536
    Specifications
    Cat. No.
    10199-506
    Concentration
  • ±0.02
    25 °C
    15 mL
    7.00
    Yellow
    Each
    Each - $22.46
    $22.46
    Each
       $22.46
     : 2283536
    Specifications
    Cat. No.
    CA10199-506
    Concentration
  • ±0.02
    25 °C
    25 mL
    7.00
    Colorless
    Each
    Each - $65.49
    $65.49
    Each
       $65.49
     : 1222220
    Specifications
    Cat. No.
    CA11017-014
    Concentration
  • ±0.02
    25 °C
    50 mL
    7.00
    Yellow
     : -
    Specifications
    Cat. No.
    10199-504
    Concentration
  • ±0.02
    25 °C
    50 mL
    7.00
    Yellow
    Each
    Each - $34.74
    $34.74
    Each
       $34.74
     : 2283526
    Specifications
    Cat. No.
    CA10199-504
    Concentration
  • ±0.02
    25 °C
    500 mL
    7.00
    Yellow
    Each (500ml)
    Each (500ml) - $52.55
    $52.55
    Each (500ml)
       $52.55
     : 2283549
    Specifications
    Cat. No.
    CA97021-458
    Concentration
  • ±0.02
    25 °C
    500 mL
    7.00
    Colorless
    Each (500ml)
    Each (500ml) - $52.07
    $52.07
    Each (500ml)
       $52.07
     : 1222249
    Specifications
    Cat. No.
    CA97021-426
    Concentration
  • ±0.02
    25 °C
    4 L
    7.00
    Yellow
    Each (4l)
    Each (4l) - $207.66
    $207.66
    Each (4l)
       $207.66
     : 2283556
    Specifications
    Cat. No.
    CA97021-440
    Concentration
  • ±0.02
    25 °C
    4 L
    7.00
    Colorless
    Each (4l)
    Each (4l) - $205.50
    $205.50
    Each (4l)
       $205.50
     : 1222256
    Specifications
    Cat. No.
    CA97021-432
    Concentration
  • ±0.02
    25 °C
    15 mL
    10.01
    Blue
     : 2283636
    Specifications
    Cat. No.
    10199-510
    Concentration
  • ±0.02
    25 °C
    15 mL
    10.01
    Blue
     : 2283636
    Specifications
    Cat. No.
    CA10199-510
    Concentration
  • ±0.02
    25 °C
    50 mL
    10.01
    Blue
     : -
    Specifications
    Cat. No.
    10199-508
    Concentration
  • ±0.02
    25 °C
    50 mL
    10.01
    Blue
    Each
    Each - $36.10
    $36.10
    Each
       $36.10
     : 2283626
    Specifications
    Cat. No.
    CA10199-508
    Concentration
  • ±0.02
    25 °C
    500 mL
    10.01
    Blue
    Each (500ml)
    Each (500ml) - $58.22
    $58.22
    Each (500ml)
       $58.22
     : 2283649
    Specifications
    Cat. No.
    CA97021-552
    Concentration
    10.01 pH ± 0.02 pH at 25 °C
  • ±0.02
    25 °C
    500 mL
    10.01
    Colorless
    Each (500ml)
    Each (500ml) - $52.25
    $52.25
    Each (500ml)
       $52.25
     : 1222149
    Specifications
    Cat. No.
    CA97021-408
    Concentration
    10.01 pH
  • ±0.02
    25 °C
    4 L
    10.01
    Blue
    Each (4l)
    Each (4l) - $174.03
    $174.03
    Each (4l)
       $174.03
     : 2283656
    Specifications
    Cat. No.
    CA22836-56
    Concentration
    10.01 ± 0.02 pH
  • ±0.02
    25 °C
    4.01
     : -
    Specifications
    Cat. No.
    CA10199-498
    Concentration
Certains produits peuvent apparaître comme restreints