Corning® Trace Elements B, Corning
Corning® Trace Elements B, Corning
Trace Elements B, 100 ml
Catalog # CA89422-908
Supplier:   Corning
Supplier #:  25-022-CI
Corning® Trace Elements B, Corning
Catalog # CA89422-908
Supplier:  Corning
CAS Number:  
Corning® Trace Elements B, Corning
Corning® Trace Elements B, Corning
Trace Elements B, 100 ml
CAS Number:  
Price
$83.80
Log in for availability
Each (100ml)
Minimum order quantity is 1
Product Details & Documents
Trace Elements B Solution has the same composition as Cleveland's Trace Element I.
The combination of Trace Elements B contains the trace elements of MCDB 104 and of MCDB 301.
This product is proprietary and not available from other major suppliers.
Specifications
Specifications
  • Désignation:
    Trace Elements B
  • Volume:
    100 ml