Plain Splinter Forceps, OR Grade, Sklar | Avantor
Plain Splinter Forceps, OR Grade, Sklar
76 mm (3"), Pointed, Serrated
Catalog # 10195-028
Supplier:   Sklar
Fournisseur #:  19-3030
Plain Splinter Forceps, OR Grade, Sklar
Catalog # 10195-028
Supplier:  Sklar
Plain Splinter Forceps, OR Grade, Sklar
Plain Splinter Forceps, OR Grade, Sklar
76 mm (3"), Pointed, Serrated
Price
$115.57
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Plain Splinter Forceps are multi-purpose thumb forceps, used for grasping and removing small foreign objects with their fine, pointed tips
They may be used for holding bandages, sutures, or other items.
Spécifications
  • Longueur:
    76 mm (3")
  • Jaw Type:
    Serrated
  • Tip Style:
    Pointed