Human Recombinant NGF beta (from E. coli) | Avantor
Human Recombinant NGF beta (from E. coli)
Supplier:   Peprotech
Human Recombinant NGF beta (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates
Human Recombinant NGF beta (from E. coli)
5 Options Available Below
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
β-NGF is a neurotrophic factor structurally related to BDNF, NT-3 and NT-4.  These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures.  β-NGF is a potent neurotrophic factor that signals through its receptor β-NGFR, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems.  β-NGF also acts as a growth and differentiation factor for B lymphocytes, and enhances B-cell survival.  The functional form of Recombinant Human β-NGF is a non-covalently-linked homodimer of two 13.5 kDa, polypeptide monomers that each contain 120 amino acids and three disulfide bonds, which are required for biological activity.Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
SSSHPIFHRG EFSVCDSVSV WVGDKTTATD IKGKEVMVLG EVNINNSVFK QYFFETKCRD PNPVDSGCRG IDSKHWNSYC TTTHTFVKAL TMDGKQAAWR FIRIDTACVC VLSRKAVR
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • 10781-774
  • 10781-776
  • 10781-778
  • 10781-780
  • 10781-782
  • Protein/peptide name:
  • NGF beta
  • NGF beta
  • NGF beta
  • NGF beta
  • NGF beta
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Animal-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin content:
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • Biological activity:
  • Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
  • Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
  • Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
  • Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
  • Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
  • Purity:
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Formulation:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
Spécifications
Product Family Options
Informations sur le produit
  • Taille
    Origin
    Availability
    Price
  • 20 µg
    US
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (20µG)
    Each (20µG) - $314.04
    $314.04
    Each (20µG)
       $314.04
    La quantité minimale de commande est de 1
    Supplier Number: AF-450-01-20UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    10781-774
    Protein/peptide name
    NGF beta
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 100 µg
    US
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100µG)
    Each (100µG) - $654.48
    $654.48
    Each (100µG)
       $654.48
    La quantité minimale de commande est de 1
    Supplier Number: AF-450-01-100UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    10781-776
    Protein/peptide name
    NGF beta
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 250 µg
    US
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (250µG)
    Each (250µG) - $1,121.65
    $1,121.65
    Each (250µG)
       $1,121.65
    La quantité minimale de commande est de 1
    Supplier Number: AF-450-01-250UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    10781-778
    Protein/peptide name
    NGF beta
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 500 µg
    US
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500µG)
    Each (500µG) - $1,914.21
    $1,914.21
    Each (500µG)
       $1,914.21
    La quantité minimale de commande est de 1
    Supplier Number: AF-450-01-500UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    10781-780
    Protein/peptide name
    NGF beta
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 1 mg
    US
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1mg)
    Each (1mg) - $3,196.25
    $3,196.25
    Each (1mg)
       $3,196.25
    La quantité minimale de commande est de 1
    Supplier Number: AF-450-01-1MG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    10781-782
    Protein/peptide name
    NGF beta
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT