Human Recombinant IL-5 (from E. coli)
About this item
IL-5 is a hematopoietic growth factor that stimulates the proliferation and activation of eosinophils. Produced by mast cells, T cells, and eosinophils, IL-5 plays an important role in inducing cell-mediated immunity against parasitic infections and certain tumors. Elevated levels of IL-5 lead to Eosinophilia, which may result in the induction of asthma and other allergic diseases. Human and murine IL-5 are cross-species reactive. Recombinant Human IL-5 is a 26.5 kDa disulfide-linked homodimeric protein containing two 116 amino acid chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
7 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
IL-5 is a hematopoietic growth factor that stimulates the proliferation and activation of eosinophils. Produced by mast cells, T cells, and eosinophils, IL-5 plays an important role in inducing cell-mediated immunity against parasitic infections and certain tumors. Elevated levels of IL-5 lead to Eosinophilia, which may result in the induction of asthma and other allergic diseases. Human and murine IL-5 are cross-species reactive. Recombinant Human IL-5 is a 26.5 kDa disulfide-linked homodimeric protein containing two 116 amino acid chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10779-630
- 10779-632
- 10779-634
- 10779-636
- 10779-638
- 10779-640
- 10779-642
- Protein/peptide name:
- IL-5
- IL-5
- IL-5
- IL-5
- IL-5
- IL-5
- IL-5
- Protein synonyms:
- Interleukin-5|EDF|TRF|BCDFII
- Interleukin-5|EDF|TRF|BCDFII
- Interleukin-5|EDF|TRF|BCDFII
- Interleukin-5|EDF|TRF|BCDFII
- Interleukin-5|EDF|TRF|BCDFII
- Interleukin-5|EDF|TRF|BCDFII
- Interleukin-5|EDF|TRF|BCDFII
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 3567 P05113
- 3567 P05113
- 3567 P05113
- 3567 P05113
- 3567 P05113
- 3567 P05113
- 3567 P05113
- Sequence:
- MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 26.5 kDa
- 26.5 kDa
- 26.5 kDa
- 26.5 kDa
- 26.5 kDa
- 26.5 kDa
- 26.5 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- TailleOriginAvailabilityPrice
- Supplier Number: 200-05-2UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10779-630Protein/peptide nameIL-5Protein synonymsInterleukin-5|EDF|TRF|BCDFIIProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3567 P05113SequenceMIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight26.5 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 200-05-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10779-632Protein/peptide nameIL-5Protein synonymsInterleukin-5|EDF|TRF|BCDFIIProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3567 P05113SequenceMIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight26.5 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 200-05-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10779-634Protein/peptide nameIL-5Protein synonymsInterleukin-5|EDF|TRF|BCDFIIProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3567 P05113SequenceMIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight26.5 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 200-05-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10779-636Protein/peptide nameIL-5Protein synonymsInterleukin-5|EDF|TRF|BCDFIIProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3567 P05113SequenceMIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight26.5 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 200-05-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10779-638Protein/peptide nameIL-5Protein synonymsInterleukin-5|EDF|TRF|BCDFIIProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3567 P05113SequenceMIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight26.5 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 200-05-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10779-640Protein/peptide nameIL-5Protein synonymsInterleukin-5|EDF|TRF|BCDFIIProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3567 P05113SequenceMIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight26.5 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 200-05-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10779-642Protein/peptide nameIL-5Protein synonymsInterleukin-5|EDF|TRF|BCDFIIProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3567 P05113SequenceMIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight26.5 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient



