Human Recombinant GDF-7 (from E. coli)
À propos de cet article
GDF-7 belongs to the TGF-β superfamily of growth and differentiation factors. It is expressed selectively by roof plate cells that are located in the developing embryonic central nervous system, and has been shown to influence the neuronal identity of cells within the central nervous system. GDF-7 has also been implicated in the formation, maintenance, and repair of certain cartilage and ligament tissues. Recombinant Human GDF-7 is a 28.0 kDa disulfide-linked homodimer containing two 129 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
6 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
GDF-7 belongs to the TGF-β superfamily of growth and differentiation factors. It is expressed selectively by roof plate cells that are located in the developing embryonic central nervous system, and has been shown to influence the neuronal identity of cells within the central nervous system. GDF-7 has also been implicated in the formation, maintenance, and repair of certain cartilage and ligament tissues. Recombinant Human GDF-7 is a 28.0 kDa disulfide-linked homodimer containing two 129 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 10779-188
- 10779-190
- 10779-192
- 10779-194
- 10779-196
- 10779-198
- Protein/peptide name:
- GDF-7
- GDF-7
- GDF-7
- GDF-7
- GDF-7
- GDF-7
- Protein synonyms:
- BMP-12|Growth/Differentiation Factor-7
- BMP-12|Growth/Differentiation Factor-7
- BMP-12|Growth/Differentiation Factor-7
- BMP-12|Growth/Differentiation Factor-7
- BMP-12|Growth/Differentiation Factor-7
- BMP-12|Growth/Differentiation Factor-7
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 151449 Q7Z4P5
- 151449 Q7Z4P5
- 151449 Q7Z4P5
- 151449 Q7Z4P5
- 151449 Q7Z4P5
- 151449 Q7Z4P5
- Sequence:
- TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
- TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
- TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
- TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
- TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
- TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 28 kDa
- 28 kDa
- 28 kDa
- 28 kDa
- 28 kDa
- 28 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Spécifications
Product Family Options
Informations sur le produit
- TailleOriginAvailabilityPrice
- Supplier Number: 120-37-2UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10779-188Protein/peptide nameGDF-7Protein synonymsBMP-12|Growth/Differentiation Factor-7Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID151449 Q7Z4P5SequenceTALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCRProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight28 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 120-37-10UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10779-190Protein/peptide nameGDF-7Protein synonymsBMP-12|Growth/Differentiation Factor-7Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID151449 Q7Z4P5SequenceTALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCRProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight28 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 120-37-100UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10779-192Protein/peptide nameGDF-7Protein synonymsBMP-12|Growth/Differentiation Factor-7Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID151449 Q7Z4P5SequenceTALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCRProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight28 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 120-37-250UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10779-194Protein/peptide nameGDF-7Protein synonymsBMP-12|Growth/Differentiation Factor-7Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID151449 Q7Z4P5SequenceTALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCRProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight28 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 120-37-500UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10779-196Protein/peptide nameGDF-7Protein synonymsBMP-12|Growth/Differentiation Factor-7Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID151449 Q7Z4P5SequenceTALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCRProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight28 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 120-37-1MGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10779-198Protein/peptide nameGDF-7Protein synonymsBMP-12|Growth/Differentiation Factor-7Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID151449 Q7Z4P5SequenceTALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCRProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight28 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



