Cytokine
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of your mouse dendritic cells.
- Animal-free
The package contains the following 2 vials:
100ug Animal-Free Murine IL-4
100ug Animal-Free Murine GM-CSF
To request a CoA or MSDS for the components of the package, please Contact Us. Manufactured using all Animal-Free reagents.
To request a CoA or MSDS for the components of the package, please Contact Us. Manufactured using all Animal-Free reagents.
Carrier free.
: For research use only. Not for use in diagnostic or therapeutic procedures.
Spécifications
Spécifications
- Taille:100 µg
- Origin:US
- Cat. No.:10778-866
- Endotoxin content:<0.1 ng/μg of protein (<1 EU/μg).
- Protein/peptide name:Cytokine
- Storage conditions:−20 °C
- Formulation:Lyophilized
- Animal-free:Y
- Shipping temperature:RT
Spécifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



