Human Recombinant IL36G (from E. coli)
À propos de cet article
The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36 α (IL-1F6), β (IL-1F8) and γ (IL-1F9). IL-36γ is highly expressed in psoriatic plaques and in tissues containing epithelial cells. IL-36γ signals through the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells. The interaction of the IL-1Rrp2 receptor with IL-36 ligands induces dendritic cell maturation and activation. IL-36γ also functions as an agonist of NF-κB, and can stimulate the inflammatory response in bronchial epithelial cells. Recombinant Human IL-36γ is a 17.0 kDa protein containing 152 amino acid residues. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
SMCKPITGTI NDLNQQVWTL QGQNLVAVPR SDSVTPVTVA VITCKYPEAL EQGRGDPIYL GIQNPEMCLY CEKVGEQPTL QLKEQKIMDL YGQPEPVKPF LFYRAKTGRT STLESVAFPD WFIASSKRDQ PIILTSELGK SYNTAFELNI ND
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
7 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36 α (IL-1F6), β (IL-1F8) and γ (IL-1F9). IL-36γ is highly expressed in psoriatic plaques and in tissues containing epithelial cells. IL-36γ signals through the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells. The interaction of the IL-1Rrp2 receptor with IL-36 ligands induces dendritic cell maturation and activation. IL-36γ also functions as an agonist of NF-κB, and can stimulate the inflammatory response in bronchial epithelial cells. Recombinant Human IL-36γ is a 17.0 kDa protein containing 152 amino acid residues. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
SMCKPITGTI NDLNQQVWTL QGQNLVAVPR SDSVTPVTVA VITCKYPEAL EQGRGDPIYL GIQNPEMCLY CEKVGEQPTL QLKEQKIMDL YGQPEPVKPF LFYRAKTGRT STLESVAFPD WFIASSKRDQ PIILTSELGK SYNTAFELNI ND
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 10778-430
- 10778-432
- 10778-434
- 10778-436
- 10778-438
- 10778-440
- 10778-442
- Protein/peptide name:
- IL36G
- IL36G
- IL36G
- IL36G
- IL36G
- IL36G
- IL36G
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Animal-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin content:
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- Biological activity:
- Measured by its ability to induce IL-8 production by human PBMCs.
- Measured by its ability to induce IL-8 production by human PBMCs.
- Measured by its ability to induce IL-8 production by human PBMCs.
- Measured by its ability to induce IL-8 production by human PBMCs.
- Measured by its ability to induce IL-8 production by human PBMCs.
- Measured by its ability to induce IL-8 production by human PBMCs.
- Measured by its ability to induce IL-8 production by human PBMCs.
- Purity:
- Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Formulation:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- RT
- RT
Spécifications
Product Family Options
Informations sur le produit
- TailleOriginAvailabilityPrice
- Supplier Number: AF-200-36G-2UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10778-430Protein/peptide nameIL36GProtein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityMeasured by its ability to induce IL-8 production by human PBMCs.PurityGreater than 95% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-36G-10UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10778-432Protein/peptide nameIL36GProtein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityMeasured by its ability to induce IL-8 production by human PBMCs.PurityGreater than 95% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-36G-50UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10778-434Protein/peptide nameIL36GProtein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityMeasured by its ability to induce IL-8 production by human PBMCs.PurityGreater than 95% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-36G-100UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10778-436Protein/peptide nameIL36GProtein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityMeasured by its ability to induce IL-8 production by human PBMCs.PurityGreater than 95% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-36G-250UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10778-438Protein/peptide nameIL36GProtein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityMeasured by its ability to induce IL-8 production by human PBMCs.PurityGreater than 95% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-36G-500UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10778-440Protein/peptide nameIL36GProtein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityMeasured by its ability to induce IL-8 production by human PBMCs.PurityGreater than 95% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-36G-1MGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10778-442Protein/peptide nameIL36GProtein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityMeasured by its ability to induce IL-8 production by human PBMCs.PurityGreater than 95% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT