Human Recombinant IL6 (from E. coli)
IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6Rα and gp130. Murine IL-6 is inactive on human cells, while both human and murine are equally active on murine cells. Recombinant Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues.Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
PVPPGEDSKD VAAPHRQPLT SSERIDKQIR YILDGISALR KETCNKSNMC ESSKEALAEN NLNLPKMAEK DGCFQSGFNE ETCLVKIITG LLEFEVYLEY LQNRFESSEE QARAVQMSTK VLIQFLQKKA KNLDAITTPD PTTNASLLTK LQAQNQWLQD MTTHLILRSF KEFLQSSLRA LRQM
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6Rα and gp130. Murine IL-6 is inactive on human cells, while both human and murine are equally active on murine cells. Recombinant Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues.Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
PVPPGEDSKD VAAPHRQPLT SSERIDKQIR YILDGISALR KETCNKSNMC ESSKEALAEN NLNLPKMAEK DGCFQSGFNE ETCLVKIITG LLEFEVYLEY LQNRFESSEE QARAVQMSTK VLIQFLQKKA KNLDAITTPD PTTNASLLTK LQAQNQWLQD MTTHLILRSF KEFLQSSLRA LRQM
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Catalog No:
- 10778-280
- 10778-282
- 10778-284
- 10778-286
- 10778-288
- 10778-290
- 10778-292
- Protein/peptide name:
- IL6
- IL6
- IL6
- IL6
- IL6
- IL6
- IL6
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Animal-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin content:
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- Biological activity:
- The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- Purity:
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- RT
- RT
Product Family Options
- TailleOrigin
- : AF-200-06-5UG
Specifications
Cat. No.10778-280Protein/peptide nameIL6Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - : AF-200-06-20UG
Specifications
Cat. No.10778-282Protein/peptide nameIL6Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - : AF-200-06-50UG
Specifications
Cat. No.10778-284Protein/peptide nameIL6Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - : AF-200-06-100UG
Specifications
Cat. No.10778-286Protein/peptide nameIL6Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - : AF-200-06-250UG
Specifications
Cat. No.10778-288Protein/peptide nameIL6Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - : AF-200-06-500UG
Specifications
Cat. No.10778-290Protein/peptide nameIL6Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - : AF-200-06-1MG
Specifications
Cat. No.10778-292Protein/peptide nameIL6Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT