Human Recombinant IL2 (from E. coli)
About this item
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant Human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFAQSII STLT
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
6 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant Human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFAQSII STLT
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10778-240
- 10778-242
- 10778-244
- 10778-246
- 10778-248
- 10778-250
- Protein/peptide name:
- IL2
- IL2
- IL2
- IL2
- IL2
- IL2
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Animal-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin content:
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- Biological activity:
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- Purity:
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- TailleOriginAvailabilityPrice
- Supplier Number: AF-200-02-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-240Protein/peptide nameIL2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-02-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-242Protein/peptide nameIL2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-02-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-244Protein/peptide nameIL2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-02-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-246Protein/peptide nameIL2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-02-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-248Protein/peptide nameIL2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT - Supplier Number: AF-200-02-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-250Protein/peptide nameIL2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliAnimal-freeYEndotoxin content<0.1 ng/μg of protein (<1 EU/μg).Biological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage conditions−20 °CShipping temperatureRT