Mouse Recombinant RANTES (from E. coli)
À propos de cet article
RANTES is a CC chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant Murine RANTES (CCL5) is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
3 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
RANTES is a CC chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant Murine RANTES (CCL5) is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 10773-172
- 10773-174
- 10773-176
- Protein/peptide name:
- RANTES
- RANTES
- RANTES
- Protein synonyms:
- SIS-delta|Regulation upon Activation Normal T-cell Express Sequence
- SIS-delta|Regulation upon Activation Normal T-cell Express Sequence
- SIS-delta|Regulation upon Activation Normal T-cell Express Sequence
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 20304 P30882
- 20304 P30882
- 20304 P30882
- Sequence:
- SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS
- SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS
- SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS
- Protease-free:
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 7.8 kDa
- 7.8 kDa
- 7.8 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
Spécifications
Product Family Options
Informations sur le produit
- TailleOriginAvailabilityPrice
- Supplier Number: 250-07-5UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10773-172Protein/peptide nameRANTESProtein synonymsSIS-delta|Regulation upon Activation Normal T-cell Express SequenceProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20304 P30882SequenceSPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMSProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 250-07-20UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10773-174Protein/peptide nameRANTESProtein synonymsSIS-delta|Regulation upon Activation Normal T-cell Express SequenceProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20304 P30882SequenceSPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMSProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - Supplier Number: 250-07-50UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10773-176Protein/peptide nameRANTESProtein synonymsSIS-delta|Regulation upon Activation Normal T-cell Express SequenceProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20304 P30882SequenceSPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMSProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



