Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant IL-8 (from <i>E. coli</i>)

Human Recombinant IL-8 (from E. coli)

1 mg, US
Catalog # 10772-954
Supplier:   Peprotech
Supplier #:  200-08-1MG
Human Recombinant IL-8 (from <i>E. coli</i>)
Catalog # 10772-954
Supplier:  Peprotech
CAS Number:  
Human Recombinant IL-8 (from E. coli)

1 mg, US

CAS Number:  
Price
$7,969.24
Log in for availability
Each (1mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

IL-8 is a proinflammatory CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. IL-8 chemoattracts and activates neutrophils. Recombinant Human IL-8 (CXCL9) (endothelial-derived) is an 8.9 kDa protein containing 77 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications

Specifications

  • Taille:
    1 mg
  • Origin:
    US
  • Protease-free:
    Y
  • Form:
    Lyophilized
  • Protein/peptide name:
    IL-8
  • Protein synonyms:
    Neutrophil Activating Factor (NAF)|Interleukin-8|NAP-1|Monocyte-Derived Neutrophil Chemotactic Factor (MDNCF)
  • Protein Function:
    Cytokines and Growth Factors
  • Purity:
    0.98
  • Endotoxin level:
    Low
  • Sequence:
    AVLPRSAKEL RCQCIKTYSK PFHPKFIKEL RVIESGPHCA NTEIIVKLSD GRELCLDPKE NWVQRVVEKF LKRAENS
  • Source:
    E. coli
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Molecular weight:
    8.9 kDa
  • Shipping temperature:
    Ambient
  • Cat. No.:
    10772-954
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    >0.1 ng/μg
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Gene ID:
    3576 P10145
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Conjugation:
    Unconjugated
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.