Human Recombinant SCGF-alpha (from E. coli)
SCGF-α and -β are hematopoietic growth factors that exert their activity at early stages of hematopoiesis. The SCGFs are non-glycosylated, species-specific cytokines that can support growth of primitive hematopoietic cells, and, in combination with EPO or GM-CSF, promote proliferation of erythroid or myeloid progenitors, respectively. Recombinant Human SCGF-α is a 33.9 kDa polypeptide containing 305 amino acid residues.
: For research use only. Not for use in diagnostic or therapeutic procedures.
SCGF-α and -β are hematopoietic growth factors that exert their activity at early stages of hematopoiesis. The SCGFs are non-glycosylated, species-specific cytokines that can support growth of primitive hematopoietic cells, and, in combination with EPO or GM-CSF, promote proliferation of erythroid or myeloid progenitors, respectively. Recombinant Human SCGF-α is a 33.9 kDa polypeptide containing 305 amino acid residues.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Catalog No:
- 10772-062
- 10772-064
- 10772-066
- 10772-068
- 10772-070
- 10772-072
- Protein/peptide name:
- SCGF-alpha
- SCGF-alpha
- SCGF-alpha
- SCGF-alpha
- SCGF-alpha
- SCGF-alpha
- Protein synonyms:
- Stem Cell Growth Factor-α
- Stem Cell Growth Factor-α
- Stem Cell Growth Factor-α
- Stem Cell Growth Factor-α
- Stem Cell Growth Factor-α
- Stem Cell Growth Factor-α
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 6320 Q9Y240
- 6320 Q9Y240
- 6320 Q9Y240
- 6320 Q9Y240
- 6320 Q9Y240
- 6320 Q9Y240
- Sequence:
- GHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPF
- GHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPF
- GHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPF
- GHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPF
- GHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPF
- GHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPF
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 33.9 kDa
- 33.9 kDa
- 33.9 kDa
- 33.9 kDa
- 33.9 kDa
- 33.9 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Product Family Options
- TailleOrigin
- : 100-22A-2UG
Specifications
Cat. No.10772-062Protein/peptide nameSCGF-alphaProtein synonymsStem Cell Growth Factor-αProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6320 Q9Y240SequenceGHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight33.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 100-22A-10UG
Specifications
Cat. No.10772-064Protein/peptide nameSCGF-alphaProtein synonymsStem Cell Growth Factor-αProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6320 Q9Y240SequenceGHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight33.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 100-22A-100UG
Specifications
Cat. No.10772-066Protein/peptide nameSCGF-alphaProtein synonymsStem Cell Growth Factor-αProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6320 Q9Y240SequenceGHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight33.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 100-22A-250UG
Specifications
Cat. No.10772-068Protein/peptide nameSCGF-alphaProtein synonymsStem Cell Growth Factor-αProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6320 Q9Y240SequenceGHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight33.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 100-22A-500UG
Specifications
Cat. No.10772-070Protein/peptide nameSCGF-alphaProtein synonymsStem Cell Growth Factor-αProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6320 Q9Y240SequenceGHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight33.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 100-22A-1MG
Specifications
Cat. No.10772-072Protein/peptide nameSCGF-alphaProtein synonymsStem Cell Growth Factor-αProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6320 Q9Y240SequenceGHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight33.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient



