Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Human Recombinant Heregulinbeta-1 (from E. coli)

 :   Peprotech
Human Recombinant Heregulinbeta-1 (from <i>E. coli</i>)
 :  Peprotech
 
Human Recombinant Heregulinbeta-1 (from E. coli)

 

 

Neuregulin/Heregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3 and NRG4).

  • Recombinant Human Heregulinβ-1 is a 7.5 kDa polypeptide consisting of only the EGF domain of heregulinβ-1 (65 amino acid residues)

To date, there are over 14 soluble and transmembrane proteins derived from the NRG1 gene. Proteolytic processing of the extracellular domain of the transmembrane NRG1 isoforms releases soluble growth factors. HRG1-β1 contains an Ig domain and an EGF-like domain; the latter is necessary for direct binding to receptor tyrosine kinases erb3 and erb4. This binding induces erb3 and erb4 heterodimerization with erb2, stimulating intrinsic kinase activity that leads to tyrosine phosphorylation. Although HRG1-β1's biological effects are still unclear, it has been found to promote motility and invasiveness of breast cancer cells, which may also involve up-regulation of expression and function of the autocrine motility-promoting factor (AMF).

 : For research use only. Not for use in diagnostic or therapeutic procedures.

 

 

 

  • Catalog No:
  • 10770-630
  • 10770-632
  • 10770-634
  • 10770-636
  • 10770-638
  • 10770-640
  • Protein/peptide name:
  • Heregulinbeta-1
  • Heregulinbeta-1
  • Heregulinbeta-1
  • Heregulinbeta-1
  • Heregulinbeta-1
  • Heregulinbeta-1
  • Protein synonyms:
  • HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
  • HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
  • HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
  • HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
  • HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
  • HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 3084 Q02297
  • 3084 Q02297
  • 3084 Q02297
  • 3084 Q02297
  • 3084 Q02297
  • 3084 Q02297
  • Sequence:
  • SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
  • SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
  • SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
  • SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
  • SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
  • SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
  • Protease-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular weight:
  • 7.5 kDa
  • 7.5 kDa
  • 7.5 kDa
  • 7.5 kDa
  • 7.5 kDa
  • 7.5 kDa
  • Reconstitution instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

 

Product Family Options

 

  • Taille
    Origin
     
     
  • 10 μg
    US
       
       
    Each (10µG)
    Each (10µG) - 243,58 $
    243,58 $
    Each (10µG)
       243,58 $
     

     : 100-03-10UG
     
     
     
    Specifications
    Cat. No.
    10770-630
    Protein/peptide name
    Heregulinbeta-1
    Protein synonyms
    HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3084 Q02297
    Sequence
    SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    7.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 50 μg
    US
       
       
    Each (50µG)
    Each (50µG) - 591,67 $
    591,67 $
    Each (50µG)
       591,67 $
     

     : 100-03-50UG
     
     
     
    Specifications
    Cat. No.
    10770-632
    Protein/peptide name
    Heregulinbeta-1
    Protein synonyms
    HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3084 Q02297
    Sequence
    SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    7.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 100 μg
    US
       
       
    Each (100µG)
    Each (100µG) - 954,95 $
    954,95 $
    Each (100µG)
       954,95 $
     

     : 100-03-100UG
     
     
     
    Specifications
    Cat. No.
    10770-634
    Protein/peptide name
    Heregulinbeta-1
    Protein synonyms
    HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3084 Q02297
    Sequence
    SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    7.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 250 μg
    US
       
       
    Each (250µG)
    Each (250µG) - 1 591,63 $
    1 591,63 $
    Each (250µG)
       1 591,63 $
     

     : 100-03-250UG
     
     
     
    Specifications
    Cat. No.
    10770-636
    Protein/peptide name
    Heregulinbeta-1
    Protein synonyms
    HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3084 Q02297
    Sequence
    SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    7.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 500 μg
    US
       
       
    Each (500µG)
    Each (500µG) - 2 518,69 $
    2 518,69 $
    Each (500µG)
       2 518,69 $
     

     : 100-03-500UG
     
     
     
    Specifications
    Cat. No.
    10770-638
    Protein/peptide name
    Heregulinbeta-1
    Protein synonyms
    HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3084 Q02297
    Sequence
    SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    7.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 1 mg
    US
       
       
    Each (1mg)
    Each (1mg) - 4 550,16 $
    4 550,16 $
    Each (1mg)
       4 550,16 $
     

     : 100-03-1MG
     
     
     
    Specifications
    Cat. No.
    10770-640
    Protein/peptide name
    Heregulinbeta-1
    Protein synonyms
    HRG1-β1|glial growth factor|ARIA (Acetylcholine Receptor Inducing Activity)|Breast cancer cell differentiation factor p45|HRG|Neuregulin1 (NRG1-β1)|Neu differentiation factor (rat)
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3084 Q02297
    Sequence
    SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    7.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.