Anti-TSPAN9 Rabbit Polyclonal Antibody (Cy3®)
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The tetraspanin family is a group of cell surface proteins that regulate cell development, activation, growth and motility. Each member contains four hydrophobic domains and participates in the mediation of signal transduction. NET-5, also known as TSPAN9 (tetraspanin 9), is a 239 amino acid multi-pass membrane protein that belongs to the tetraspanin (TM4SF) family. NET-5 forms a complex with GPVI in the tetraspanin microdomains on the platelet surface, and is encoded by a gene that maps to human chromosome 12p13.33. Chromosome 12 encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSPAN9
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Spécifications
- Taille:100 µL
- Clonality:Polyclonal
- Flow cytometry:Yes
- Purification:Purified by Protein A
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Store at –20 °C for 12 months
- Host:Rabbit
- Concentration:1 μg/μl
- Cross adsorption:No
- Shipping temperature:4°C
- Antigen name:Teraspanin 9
- Cat. No.:10490-004
- Reactivity:Human|Rat|Mouse
- Gene ID:10867
- Antigen synonyms:NET5|PP1057|Tspan-9|NET-5|TSPAN9|Tetraspan NET-5|Tetraspanin-9
- Immunogen:180-203/239
- ImmunoChemistry:Yes
- Conjugation:Cy3®
- Antigen symbol:TSPAN9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.



