Anti-HIPK3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10454-502
: Bioss
- Antibody type:Primary
- Antigen name:HomeoDomain Interacting Protein kinase 3
- Antigen symbol:HIPK3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Gene ID:10114
- Antigen synonyms:Fas-interacting serine/threonine-protein kinase|ANPK|Homolog of protein kinase YAK1|DYRK6|Homeodomain-interacting protein kinase 3|Androgen receptor-interacting nuclear protein kinase|FIST|PKY|FIST3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10454-502
- Supplier No.:BS-6777R-FITC
Seems to negatively regulate apoptosis by promoting FADD phosphorylation. Enhances androgen receptor-mediated transcription. May act as a transcriptional corepressor for NK homeodomain transcription factors.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HIPK3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat