Spécifications
- Antibody type:Primary
- Antigen symbol:PCANAP2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Gene ID:85414
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10450-034
- Supplier No.:BS-6568R-FITC
Spécifications
À propos de cet article
PSA, prostate specific antigen, is the classic indicator for transformed pro-state tissue; however, in addition to being upregulated in prostate cancer, PSA is also upregulated in non-malignant conditions, such as benign prostatic hyperplasia. Prostein, also designated Prostate cancer-associated protein 6, is a prostate-specific, 553 amino acid transmembrane protein that is upregulated by androgens. It is considered a marker for prostate cells since it is expressed in all prostatic glandular cells as well as in normal and cancerous prostate tissues. Since it is able to elicit a tumor-directed cytotoxic T cell response, Prostein may be used as a target for the development of PSA- and T cell-based therapeutic strategies for prostate cancer.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PCANAP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat