Anti-LDOC1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10449-432
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Leucine zipper protein down-regulated in cancer cells
- Antigen symbol:LDOC1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Gene ID:23641
- Antigen synonyms:Leucine zipper protein down-regulated in cancer cells|Protein LDOC1|LDOC1|Mar7|BCUR1|Mart7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:100-146/146
- Purification:Purified by Protein A
- Cat. no.:10449-432
- Supplier No.:BS-6543R-CY3
Specifications
About this item
May have an important role in the development and/or progression of some cancers.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LDOC1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human