Anti-KRV-VP1/KRV-VP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10398-354
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Kilham Rat Virus
- Antigen symbol:KRV-VP1/KRV-VP2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:KRV-VP1/2|viral protein 2.|viral protein 2|viral protein 1|viral protein 1/2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:515-565/582
- Purification:Purified by Protein A
- Cat. no.:10398-354
- Supplier No.:BS-4642R-FITC
Specifications
About this item
Kilham Rat Virus/KRV-VP1/KRV-VP2 (C-terminus)
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KRV-VP1/KRV-VP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus