Anti-CDKN2A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10398-224
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase Inhibitor 2A
- Antigen symbol:CDKN2A
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Gene ID:1029
- Antigen synonyms:CDKN2|MLM|INK4|MTS-1|P16|P16INK4|P19|P16-INK4A|P16INK4A|INK4A|P19ARF|CDK4I|CMM2|P14ARF|P14|ARF|MTS1|TP16
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:20-70/156
- Purification:Purified by Protein A
- Cat. no.:10398-224
- Supplier No.:BS-4592R-FITC
Specifications
About this item
Acts as a negative regulator of the proliferation of normal cells by interacting strongly with CDK4 and CDK6. This inhibits their ability to interact with cyclins D and to phosphorylate the retinoblastoma protein.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDKN2A
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: