Spécifications
- Antibody type:Primary
- Antigen name:CSFV Envelope glycoprotein E2
- Antigen symbol:CSFV E2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:polyprotein [Classical swine fever virus]|Envelope glycoprotein E2|E2|polyprotein|structural protein E2 [Classical swine fever virus]|CSFV polyprotein|POLG_CSFVA.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:970-1020/3898
- Purification:Purified by Protein A
- Cat. no.:10398-100
- Supplier No.:BS-4527R-HRP
Spécifications
À propos de cet article
Classical swine fever(CSF) is a kind of acute and intense contagious disease in swine, it material damages swine industry. CSF could cause swine appearing disseminated hemorrhage in systemic organ and tissue, degeneration and necrosis in small vascular and capillary endothelial cells. Classical swine fever virus (CSFV) belongs to genera Pestivirus, family Flaviviridae, it’s genome total length is 12.3Kb,including a big ORF, encoding 11 structural and nonstructural proteins.Envelope glycoprotein E2 exists in the form of homodimer; this protein is cleaved from the polyprotein by a cellular signal peptidase.
Recommended Dilutions: IHC-P: 1:100-500
Type: Primary
Antigen: CSFV Envelope glycoprotein E2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: