HAEMO-SOL® Enzyme Active, Detergent | Avantor
HAEMO-SOL® Enzyme Active, Detergent
Supplier:   Haemo-Sol
HAEMO-SOL® Enzyme Active, Detergent
Supplier:  Haemo-Sol
Certificates
HAEMO-SOL® Enzyme Active, Detergent
HAEMO-SOL® Enzyme Active, Detergent
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Concentrated enzymatic detergent for hard to remove protein wastes. Breaks down dried, heavy or hardened soils for easy removal after soaking.
Mix 1/2 ounce to 1 gallon of warm water. Soak instruments 20 minutes or more if heavliy soiled. Use a soft brush if necessary. 5 lb makes 160 gallons of solution.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Packaging
    Availability
    Price
  • Enzymatic Detergent
    5 lb
    Jar
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $138.89
    $138.89
    Log in pour la disponibilité
    Case of 6 (1/ Itm/Bx)
    Case of 6 (1/ Itm/Bx) - $833.34
    $833.34
    Each(1/Itm/Bx)
       $138.89
    Case of 6 (1/ Itm/Bx)
       $833.34
    Supplier Number: 026-055
  • 30× 0.5 oz
    Packets
    Discontinued
    Supplier Number: -