Polyester Printer Labels, Brady
Polyester Printer Labels, Brady
 :   Avantor
Polyester Printer Labels, Brady
 :  Avantor
Polyester Printer Labels, Brady
Polyester Printer Labels, Brady
B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.
B-498: Vinyl cloth backing offers excellent holding power and offers clean removal and repositioning.
B-424: Paper labels with permanent rubber adhesive provide good contrast and smear resistance for thermal transfer and barcode printing applications.
B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-430: Clear polyester labels are designed for applications that use barcodes, alphanumeric, graphic symbols and logos and require nameplate-like quality.
B-499: Nylon cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
B-351: Tamper resistant vinyl is designed to fracture easily to show signs of product tampering and to prevent on-piece label removal.
B-423: Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-439: Permanent vinyl is a thermal transfer printable material ideal for applications needing various colors like finished product, rating plates and general-purpose printers.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
Product Family Options
B-461 Ultra-Thin Label with Clear Tail for Tubes
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-461 MAT´L
    Polyester
    Yellow/translucent
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    2.625×1"
    Each
    Each - $55.25
    $55.25
    Each
       $55.25
B-490 Freezerbondz® Label for Frozen Tubes
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-490 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1.5×3.875"
    Each
    Each - $97.84
    $97.84
    Each
       $97.84
B-498 Tissue Cassette Label for Reidentification of Processed Paraffin Blocks
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-498 MAT´L
    Vinyl Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×1"
    Each
    Each - $114.52
    $114.52
    Each
       $114.52
  • Tls printer labels B-498 MAT´L
    Vinyl Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×1.5"
    Each
    Each - $101.17
    $101.17
    Each
       $101.17
  • Tls printer labels B-498 MAT´L
    Vinyl Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.75×0.5"
    Each
    Each - $85.59
    $85.59
    Each
       $85.59
  • Tls printer labels B-498 MAT´L
    Vinyl Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1×0.375"
    Each
    Each - $108.96
    $108.96
    Each
       $108.96
  • Tls printer labels B-498 MAT´L
    Vinyl Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1.5×1"
    Each
    Each - $96.71
    $96.71
    Each
       $96.71
B-424 Economical Paper Label for General Purpose
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-424 MAT´L
    Paper
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.9×0.75"
    Each
    Each - $38.35
    $38.35
    Each
       $38.35
B-427 Straw Label for Wrapping
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-427 MAT´L
    Vinyl
    White/translucent
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×2.2"
    Each
    Each - $32.01
    $32.01
    Each
       $32.01
  • Tls printer labels B-427 MAT´L
    Vinyl
    White/translucent
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    2.5×1"
    Each
    Each - $50.03
    $50.03
    Each
       $50.03
B-430 Petri Dish and Bag Label
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-430 MAT´L
    Polyester
    Clear
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×1"
    Each
    Each - $95.61
    $95.61
    Each
       $95.61
B-499 For Cryo Boxes, Freezer Racks, Petri Dishes, Bottles, Flasks, Microplates, and Bags
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-499 MAT´L
    Nylon Cloth
    Yellow
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.375"
    Each
    Each - $66.70
    $66.70
    Each
       $66.70
  • Tls printer labels B-499 MAT´L
    Nylon Cloth
    Yellow
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5"
    Each
    Each - $60.58
    $60.58
    Each
       $60.58
  • Tls printer labels B-499 MAT´L
    Nylon Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.25×1.9"
    Each
    Each - $117.85
    $117.85
    Each
       $117.85
  • Tls printer labels
    Nylon Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.375×1"
    Each
    Each - $123.02
    $123.02
    Each
       $123.02
  • Tls printer labels
    Nylon Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×0.9"
    Each
    Each - $95.15
    $95.15
    Each
       $95.15
  • Tls printer labels B-499 MAT´L
    Nylon Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1×0.375"
    Each
    Each - $111.17
    $111.17
    Each
       $111.17
  • Tls printer labels B-499 MAT´L
    Nylon Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1.5×0.75"
    Each
    Each - $90.05
    $90.05
    Each
       $90.05
  • Tls printer labels B-499 MAT´L
    Nylon Cloth
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1.5×1"
    Each
    Each - $104.50
    $104.50
    Each
       $104.50
B-351 Removes in Fragments, Good Resistance to Solvents and Humidity, UL and RoHS
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-351 MAT´L
    Vinyl
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×1"
    Each
    Each - $108.96
    $108.96
    Each
       $108.96
  • Tls printer labels B-351 MAT´L
    Vinyl
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1×2"
    Each
    Each - $65.59
    $65.59
    Each
       $65.59
  • Tls printer labels B-351 MAT´L
    Vinyl
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1.9×3"
    Each
    Each - $96.71
    $96.71
    Each
       $96.71
B-423 Product and Component Identification Labels, PCB and Rating Plates
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.19×1"
    Each
    Each - $101.17
    $101.17
    Each
       $101.17
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.25×2"
    Each
    Each - $35.35
    $35.35
    Each
       $35.35
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.275×1"
    Each
    Each - $103.40
    $103.40
    Each
       $103.40
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×0.5"
    Each
    Each - $71.15
    $71.15
    Each
       $71.15
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    0.5×1"
    Each
    Each - $81.17
    $81.17
    Each
       $81.17
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1×1.5"
    Each
    Each - $63.36
    $63.36
    Each
       $63.36
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1×2"
    Each
    Each - $45.90
    $45.90
    Each
       $45.90
  • Tls printer labels B-423 MAT´L
    Polyester
    White
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
    1×3"
    Each
    Each - $56.70
    $56.70
    Each
       $56.70
Other BMP®71 Labels
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Width
    Dimensions
    Roll Length
  • Tls print ribbon blue
    Blue
    TLS 2200®, TLS PC Link®, BMP®71
    2"
    75 ft.
  • Ribbons TT4300SER
    Black
    BMP®71
    2"
    150 ft.
    Each
    Each - $95.17
    $95.17
    Each
       $95.17
  • Polyester printer labels
    Black
    BMP®71
    2"
    150 ft.