Robust, universal insulated shipping solutions optimized to provide the lowest total cost of ownership available.
- One-way universal solutions
- Operational efficiency, redefined
- Material neutral approach
- PureTemp® vegetable-based phase change material
Our Certis line of premium pre-qualifed insulated shippers are ideal for one-way shipments of pharmaceutical products across summer or winter seasons for refrigerated, CRT and frozen shipping solutions from 48 hours to 120 hours of duration.
Material neutral approach utilizes leading edge technologies to provide the lowest total cost of ownership. Designed solutions reduce the amount of weight, packout complexity, and overall size, resulting in reduced shipping costs. Designs feature PureTemp® Vegetable-Based Phase Change Material, which is a natural phase change material (PCM) that provides strict temperature control and greater efficiency, resulting in packages that are smaller and lighter.
The Certis Platform of shippers are designed to offer operational efficiency and ease of personnel training using ZERO Bench-Time® technology and same pack-out configuration across durations. Certis Platform utilizes the same pack-out for refrigerated, CRT and frozen solutions from 48 hours to 120 hours of duration. These shippers optimize inventory and provide lowest total cost of ownership utilizing only 3 types of bricks and 8 shippers to create 24 shipping solutions across temperature ranges and duration.
Designed to a variety of different ambient temperature profiles representative of moderate to extreme external distribution environments.