Innovating Science® Document Analysis: Comprehensive Lab Activities
À propos de cet article
This kit has been designed to introduce students to the field of document analysis. Experiments in this kit cover the areas of paper analysis, hand writing analysis, detection of a forgery, recovery of mechanical/chemical erasures, hidden message discovery and ink analysis. Students will make use of techniques like microscopy, chromatography and fuming to complete the activities. Kit contains 10 unique sets of materials for 10 groups. Teacher’s Manual and Student Study Guide copymasters are included.
- Kit contains 10 unique sets of materials for 10 groups
- Teacher’s Manual and Student Study Guide copymasters are included
Experiments in this kit cover the areas of paper analysis, hand writing analysis, detection of a forgery, recovery of mechanical/chemical erasures, hidden message discovery and ink analysis. Students will make use of techniques like microscopy, chromatography and fuming to complete the activities.
2 Options Available Below
Product Details & Documents
This kit has been designed to introduce students to the field of document analysis. Experiments in this kit cover the areas of paper analysis, hand writing analysis, detection of a forgery, recovery of mechanical/chemical erasures, hidden message discovery and ink analysis. Students will make use of techniques like microscopy, chromatography and fuming to complete the activities. Kit contains 10 unique sets of materials for 10 groups. Teacher’s Manual and Student Study Guide copymasters are included.
- Kit contains 10 unique sets of materials for 10 groups
- Teacher’s Manual and Student Study Guide copymasters are included
Experiments in this kit cover the areas of paper analysis, hand writing analysis, detection of a forgery, recovery of mechanical/chemical erasures, hidden message discovery and ink analysis. Students will make use of techniques like microscopy, chromatography and fuming to complete the activities.