Innovating Science® Element Groups
Innovating Science® Element Groups
Element Groups, Teacher ‘s Guide and Student Copymasters, 8
Catalog # 470219-084
Supplier:   Ward's Science
Supplier #:  IS2516
Innovating Science® Element Groups
Catalog # 470219-084
Supplier:  Ward's Science
Innovating Science® Element Groups
Innovating Science® Element Groups
Element Groups, Teacher ‘s Guide and Student Copymasters, 8
Price
$284.00
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The activities in this kit focus on developing a better understanding of why periodic trends occur in the elements.
  • Kit teaches students to look for patterns as they conduct experiments
  • Students learn why the periodic table looks as it does
  • Graduated plastic pipette included
  • Two sets of experiments
Students perform two sets of experiments focusing on the alkaline earth metals and the halogens groups. In the first set of experiments, students search for patterns as they look at the reactivity of the alkaline earth metals.
The second set of experiments form small amounts of solubilized halogen gasses as a colorful way of measuring reactivity within the halogen group.
Ordering information: This kit includes ammonium oxalate, barium nitrate, calcium nitrate, chlorine water, iodine water, magnesium nitrate, mineral oil, potassium bromate, potassium bromide, potassium bromide, potassium iodate, sodium carbonate, potassium chloride, potassium iodide, strontium nitrate, sulfuric acid, sulfuric acid, graduated plastic pipette, and vials and labels for bromine water.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Spécifications
  • Désignation:
    Element Groups
  • No. of Lab Groups:
    8
  • Includes:
    Teacher ‘s Guide and Student Copymasters