ADC® Pocket Ophthalmoscope Set
470119-692
- Description:Pocket Ophthalmoscope Set
- Cat. no.:470119-692
- Supplier No.:W51514
This pocket ophthalmoscope set features a 2.5 V halogen light source interchangeable “AA” metal battery handle.
- Impact resistant head
Pocket clip with integral on/off slide switch for ambidextrous use. Lifetime quality performance guarantee on optics.