ContactUsLinkComponent
Promega® PCR Master Mix (2x)*
Catalogue # 470203-876
Numéro CAS:  
undefined
Promega® PCR Master Mix (2x)*
Catalogue # 470203-876
Supplier Number:  M7502
Numéro CAS:  
Spécifications
  • Description:
    Promega® PCR Master Mix (2x)*
  • Cat. no.:
    470203-876
  • Supplier No.:
    M7502
Spécifications
À propos de cet article
The premixed ready-to-use solution contains Taq DNA polymerase, dNTPs, and magnesium chloride at optimal concentrations for efficient amplification of DNA templates by PCR. Enough for 100 reactions (2.5 mL).
Note: Can be stored at 4°C for up to six weeks or -20° C for longer.
The premixed ready-to-use solution contains Taq DNA polymerase, dNTPs, and magnesium chloride at optimal concentrations for efficient amplification of DNA templates by PCR
Enough for 100 reactions (2.5 mL)