Iris Scissors, OR Grade, Sklar
Iris Scissors, OR Grade, Sklar
Supplier:   Sklar
Iris Scissors, OR Grade, Sklar
Supplier:  Sklar
Certificates
Iris Scissors, OR Grade, Sklar
Iris Scissors, OR Grade, Sklar
19 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris Scissors are designed for ophthalmic procedures, these fine and sharp scissors are suitable for detailed dissection of fine tissue and fine suture removal.
  • Multipurpose instrument used in various surgical environments
In addition to ophthalmic procedures, Iris scissors are used in emergency rooms and for obstetrics and gynecology as well as dermatologic procedures.
The Ready OR Grade Iris Scissors are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Product Family Options
Iris Scissors, OR Grade
Product Information
  • Désignation
    Longueur
    Matière
    Blade Type
    Curvature
    Handle
    Sterility
    Availability
    Price
  • Iris Scissor, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Angled
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $256.16
    $256.16
    New Customer Promotional Price
    Each
       $256.16
    Supplier Number: 64-2142
  • Iris Scissor, Premium OR-Grade
    8.9 cm (3.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $177.05
    $177.05
    New Customer Promotional Price
    Each
       $177.05
    Supplier Number: 47-1235
  • Iris Scissor, Premium OR-Grade
    8.9 cm (3.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $170.17
    $170.17
    New Customer Promotional Price
    Each
       $170.17
    Supplier Number: 47-1240
  • Iris Scissor, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $170.27
    $170.27
    New Customer Promotional Price
    Each
       $170.27
    Supplier Number: 47-1243
  • Iris Scissor, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $163.56
    $163.56
    New Customer Promotional Price
    Each
       $163.56
    Supplier Number: 47-1244
  • Iris Scissor, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $205.58
    $205.58
    New Customer Promotional Price
    Each
       $205.58
    Supplier Number: 47-1245
  • Iris Scissor, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved (Sideways)
    Finger Rings
    Non sterile
    Discontinued
    Supplier Number: -
  • Iris Scissor, Premium OR-Grade
    8.9 cm (3.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $180.45
    $180.45
    New Customer Promotional Price
    Each
       $180.45
    Supplier Number: 47-1135
  • Iris Scissor, Premium OR-Grade
    8.9 cm (3.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $139.13
    $139.13
    New Customer Promotional Price
    Each
       $139.13
    Supplier Number: 47-1140
  • Iris Scissor, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $161.46
    $161.46
    New Customer Promotional Price
    Each
       $161.46
    Supplier Number: 47-1143
  • Iris Scissor, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $126.46
    $126.46
    New Customer Promotional Price
    Each
       $126.46
    Supplier Number: 47-1144
  • Iris Scissor, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $139.57
    $139.57
    New Customer Promotional Price
    Each
       $139.57
    Supplier Number: 47-1145
  • Iris Scissor, 2 Probe Points, Premium OR-Grade
    10.8 cm (4.25")
    Stainless steel
    Blunt/Blunt, Smooth
    Angled
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $264.11
    $264.11
    New Customer Promotional Price
    Each
       $264.11
    Supplier Number: 64-2042
  • Iris Scissors, Delicate, Premium OR-Grade
    8.9 cm (3.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $268.23
    $268.23
    New Customer Promotional Price
    Each
       $268.23
    Supplier Number: 22-1235
  • Iris Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $281.41
    $281.41
    New Customer Promotional Price
    Each
       $281.41
    Supplier Number: 47-1248
  • Iris Scissors, Delicate, Premium OR-Grade
    8.9 cm (3.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $257.04
    $257.04
    New Customer Promotional Price
    Each
       $257.04
    Supplier Number: 22-1135
  • Iris Scissors, Delicate, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $156.94
    $156.94
    New Customer Promotional Price
    Each
       $156.94
    Supplier Number: 47-1440
  • Iris Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $343.83
    $343.83
    New Customer Promotional Price
    Each
       $343.83
    Supplier Number: 47-1246
  • Iris Scissors, Heavy, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $307.48
    $307.48
    New Customer Promotional Price
    Each
       $307.48
    Supplier Number: 47-1442
  • Iris Scissors, Heavy, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $172.26
    $172.26
    New Customer Promotional Price
    Each
       $172.26
    Supplier Number: 47-1441