3D Printer Cleaning Kit
3D Printer Cleaning Kit
Catalog # 470218-905
Supplier: DR INSTRUMENTS INC. SE
Fournisseur #: DR3DTK
Jump to:
Product Details & Documents
3D Printer Cleaning Kit contains useful tools for cleaning up 3D printed objects.
- Eight Useful Tools
- Included Bag Rolls up for Storage
- File Styles: Flat, Triangular, and Semicircular
This handy set includes eight tools to assist with a new 3D printed creation.
File styles are flat, triangular, and semicircular, and they help with cleaning up the finished prototype. The provided bag rolls up for easy storage.
: The kit is supplied with two spatulas (one wide and one narrow), three files, one metal probe, one pair of needle nose pliers, and one pair of specially designed trimming pliers.
Spécifications
- Désignation:3D Printer Cleaning Kit