About this item
The patient control slide is suitable for staining protocols that require a control.
- Spaces for Both Patient Sample and Control Material
- Chemical Adhesive for Better Tissue Adhesion
- Red Line on Reverse Side of Slide for Unrestricted Movement of Reagents
Each slide features space for both patient sample and control material
Slide is positively charged for better tissue adhesions. Available without clipped corners or with clipped corners for use in automated systems.
The slide is positively charged for better tissue adhesions. Available without clipped corners or with clipped corners for use in automated systems.
A red line allows for the two samples to be separated. This red line is printed on reverse side of slide to allow for unrestricted movement of reagents.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.