Motic BA310 Polarizing Microscope | Avantor
Motic BA310 Polarizing Microscope
Supplier:   Motic
Motic BA310 Polarizing Microscope
Supplier:  Motic
Certificates
Motic BA310 Polarizing Microscope
2 Options Available Below
Product Details & Documents
To meet increasing demands for a dedicated, professional polarizing microscope, Motic introduced the BA310Pol for observation of birefringent samples with efficiency and reliability
The BA310Pol comes standard with Motic’s high quality EC Plan objectives (4x, 10x, 40x S, 60x S) set in a reversed quadruple nosepiece and high-eyepoint N-WF PL10x widefield eyepieces for superb image clarity and field diameter. The hard-coated rotating stage features ball bearings for smooth rotation through 360°, with 1° graduations for quick, precise orientation and reference. The BA310Pol is a versatile system suited for a wide range of petrography, minerology, and material science applications where affordability and ease-of-use are key requirements.
The strain-free CCIS optics, built-up around a sturdy frame with centerable objectives, provide reproducible, high fidelity images. Full polarization is provided through the focusable and centerable Bertrand lens, 360° graduated analyzer for optimum image extinction, and a DIN-compliant compensator slot that accepts all established compensators.
Ordering information: Microscope is supplied with all accessories listed above and micrometer reticles. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA10769-628
  • CA10769-630
  • Eyepieces:
  • WF 10×/20 mm
  • WF 10×/20 mm
  • Objectives:
  • CCIS EC plan achromatic EC PL 4×, 10×, 40× S, 60× S
  • CCIS EC plan achromatic EC PL 4×, 10×, 40× S, 60× S
  • Magnification:
  • 40x to 600x
  • 40x to 600x
  • Condenser:
  • Achromat Swing-Out Condenser (NA = 0.9/0.13)
  • Achromat Swing-Out Condenser (NA = 0.9/0.13)
  • Illumination:
  • Halogen
  • Halogen
  • Optical design:
  • Infinity Corrected
  • Infinity Corrected
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • BA310Pol Binocular Polarizing Microscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,507.99
    $8,507.99
    Each
       $8,507.99
    La quantité minimale de commande est de 1
    Supplier Number: 1100100402241
    Spécifications:
    More
    Specifications
    Cat. No.
    CA10769-628
    Eyepieces
    WF 10×/20 mm
    Objectives
    CCIS EC plan achromatic EC PL 4×, 10×, 40× S, 60× S
    Magnification
    40x to 600x
    Condenser
    Achromat Swing-Out Condenser (NA = 0.9/0.13)
    Illumination
    Halogen
    Optical design
    Infinity Corrected
  • BA310Pol Trinocular Polarizing Microscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,874.24
    $8,874.24
    Each
       $8,874.24
    La quantité minimale de commande est de 1
    Supplier Number: 1100100402251
    Spécifications:
    More
    Specifications
    Cat. No.
    CA10769-630
    Eyepieces
    WF 10×/20 mm
    Objectives
    CCIS EC plan achromatic EC PL 4×, 10×, 40× S, 60× S
    Magnification
    40x to 600x
    Condenser
    Achromat Swing-Out Condenser (NA = 0.9/0.13)
    Illumination
    Halogen
    Optical design
    Infinity Corrected