JACKSON SAFETY® V80 MONOGOGGLE™ XTR OTG Goggle, KIMBERLY-CLARK PROFESSIONAL®
CA10672-498
- Frame Color:Blue
- Cat. no.:CA10672-498
- Lens Coating:Anti-fog
- Lens Tint:Clear
- Supplier No.:18624
The JACKSON SAFETY® V80 Monogoggle XTR OTG Goggle protects against splashes, while providing increased visibility due to its cylindrical lens.
A soft, pliable frame conforms to the face for comfort and is easy to clean. Indirect ventilation.
Can be worn over most prescription eyeglasses.
: ANSI Z87.1-2010 Standard.