PVC Dropper Tip, Wheaton
23 Dia. x 44 H mm, PVC, Clear
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 224080
Jump to:
Product Details & Documents
This PVC Dropper Tip attaches easily to serum bottles with a 20mm O.D
finish. The dropper tip is made from PVC, with a PE cap. It dispenses a 40μL drop using distilled water. This is an easy and effective method of converting a serum bottle to a dropping bottle.
Spécifications
- Color:Clear
- Matière:PVC
- Taille:23 Dia. x 44 H mm