Ward's® Laser Pointer
Wards laser pointer, 500 yds.(650 nm), 5mW
Catalog # 470218-644
Supplier: AIA / DICK STEIN ADVERTISING
Fournisseur #: LF1000
Jump to:
Product Details & Documents
Ward's laser pointer is perfect as a presentation aid and for teaching laser optics.
- Presentation aid
- Ideal for teaching laser optics
- Class IIIa red laser
- Compact, keychain-style
- Also features an LED flashlight
The compact laser pointer features an LED flashlight and a key chain. Included (not installed) are three long-life LR41 lithium batteries.
Three life-long LR41 lithium batteries are included. To install the batteries, remove them and the non-conductive spacer from the laser pointer, then replace the batteries.
Certifications: FDA and CE approved.
Spécifications
- Désignation:Wards laser pointer
- Range:500 yds.(650 nm), 5mW