Microtest Manifolds for Self-Refilling Syringes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Ideal for use with 24- or 96-well microplates
Tightly fitting Luer and Luer Lock syringe nozzles. Available in both a straight "T" style and an angular style with an offset hub. Stainless steel with PTFE stoppers. Autoclavable. Increased efficiency for inoculations, cell feedings, and washing.
Frequently Bought Together