PASS/FAIL® Class 5 Process Challenge Pack, Propper Manufacturing | Avantor
PASS/FAIL® Class 5 Process Challenge Pack, Propper Manufacturing
PASS/FAIL® Challenge Pack
Catalog # 10798-790
Supplier:   Propper
Fournisseur #:  26906200
PASS/FAIL® Class 5 Process Challenge Pack, Propper Manufacturing
Catalog # 10798-790
Supplier:  Propper
PASS/FAIL® Class 5 Process Challenge Pack, Propper Manufacturing
PASS/FAIL® Class 5 Process Challenge Pack, Propper Manufacturing
PASS/FAIL® Challenge Pack
Price
$832.13
Log in pour la disponibilité
Case of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PASS/FAIL® challenge pack is a disposable test pack for use in steam sterilizers.
  • Disposable test pack containing a chemical integrator
  • Provides challenge to steam sterilization loads
  • Use for load release and monitoring
PASS/FAIL® Challenge This pack contains Vapor Line® chemical integrator and is equivalent in performance to biological indicators. Integrator provides a clear, easy-to-read outcome when the indicator dark brown/black bar migrates along a paper wick from FAIL to PASS region.
This challenge pack is a disposable test pack designed to provide a challenge to steam sterilizers at 250 °F (121 °C) to 273 °C (134 °C).
Spécifications
  • Désignation:
    PASS/FAIL® Challenge Pack