Buck Neurological Hammer
Catalog # 470119-650
Supplier: American 3B Scientific
Specifications
- Description:Buck Neurological Hammer
- Length:19 cm (7½")
- Cat. no.:470119-650
- Supplier No.:W54086
Specifications
About this item
A Standard For Sensation Evaluation.
- Double-Sided Percussion Head
- Hollow Shaft Contains Point and Brush
The hammer's hollow shaft contains a removable point and brush used for cutaneous sensation evaluations.